Copper 99,9985 Metals basis, wire, Puratronic®, Ø 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
10972.G6 10972.H2
10972.G6EA 140 EUR
10972.G6 10972.H2
Copper 99,9985 Metals basis, wire, Puratronic®, Ø 0.25 mm (0.01 in)
Copper
Purity: 99.9985% CAS No.: 7440-50-8 Physical Form: ≈0.44g/m Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545