Copper 99,99 Metals basis, slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
39674.18 39674.30
39674.18EA 89.9 EUR
39674.18 39674.30
Copper 99,99 Metals basis, slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Copper
Purity: 99.99% CAS No.: 7440-50-8 Physical Form: ≈1.8g/slug Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545