Copper 99,995 Metals basis, slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
42352.09 42352.18
42352.09EA 49.5 EUR
42352.09 42352.18
Copper 99,995 Metals basis, slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Copper
Purity: 99.995% CAS No.: 7440-50-8 Physical Form: ≈0.22g/slug Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545