Copper ≥99.9999% (metals basis), wire, Puratronic®, Ø 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
42977.G6 42977.H2
42977.G6EA 193 EUR
42977.G6 42977.H2
Copper ≥99.9999% (metals basis), wire, Puratronic®, Ø 0.1 mm (0.004 in)
Copper
Purity: 99.9999% CAS No.: 7440-50-8 Physical Form: ≈0.07g/m Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545