Copper 99,99 Metals basis, wire, Ø 0.015 mm (0.0006 in)
Supplier: Thermo Fisher Scientific
43238.G6 43238.H2
43238.G6EA 220 EUR
43238.G6 43238.H2
Copper 99,99 Metals basis, wire, Ø 0.015 mm (0.0006 in)
Copper
Purity: 99.99% CAS No.: 7440-50-8 Physical Form: ≈0.0016g/m Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545