Copper ≥99.99% (metals basis), rods, Ø 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
43973.BSEA 54.7 EUR
43973.BS 43973.BV
Copper ≥99.99% (metals basis), rods, Ø 2.0 mm (0.08 in)
Copper
Purity: 99.99% CAS No.: 7440-50-8 Physical Form: ≈2.81g/10cm Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545