Zinc ≥99.99% (metals basis), rods, Ø 1.27 cm (0.5 in)
Supplier: Thermo Fisher Scientific
10433.BV 10433.RM 10433.RS
10433.BVEA 133 EUR
10433.BV 10433.RM 10433.RS
Zinc ≥99.99% (metals basis), rods, Ø 1.27 cm (0.5 in)
Zinc
Purity: 99.99+% CAS No.: 7440-66-6 Physical Form: ≈9.03g/cm Odour: Odourless MDL No.: MFCD00011291
Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255