Zinc 99 Metals basis, foil, Thickness: 1.6 mm (0.063 in)
Supplier: Thermo Fisher Scientific
10438.KG 10438.KJ
10438.KGEA 48.9 EUR
10438.KG 10438.KJ
Zinc 99 Metals basis, foil, Thickness: 1.6 mm (0.063 in)
Zinc
Purity: 99% CAS No.: 7440-66-6 Physical Form: ≈255g/15x15cm Odour: Odourless MDL No.: MFCD00011291
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255