Zinc 99,95 Metals basis, wire, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
42704.G5 42704.G9
42704.G5EA 85.7 EUR
42704.G5 42704.G9
Zinc 99,95 Metals basis, wire, Ø 0.5 mm (0.02 in)
Zinc
Purity: 99.95% CAS No.: 7440-66-6 Physical Form: ≈1.40g/m Odour: Odourless MDL No.: MFCD00011291
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255