Zinc 99,9999 Metals basis, rods, Ø 8 mm (0.31 in)
Supplier: Thermo Fisher Scientific
44936.DT 44936.DJ
44936.DTEA 892 EUR
44936.DT 44936.DJ
Zinc 99,9999 Metals basis, rods, Ø 8 mm (0.31 in)
Zinc
Purity: 99.9999% CAS No.: 7440-66-6 Odour: Odourless MDL No.: MFCD00011291
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255