Precision balances, Explorer®
Supplier: OHAUS
83021348 83021341 83021343 83021366 83021350 83021351 83021356 83021357 83021358 83021360 83021361 83021362 83021363 83021365 83021342 30061979 30061980 30061981 30061982 30061983 30061984
611-2401EA 4950 EUR
611-2401 611-2398 611-2400 611-2412 611-2402 611-2403 611-2404 611-2405 611-2406 611-2407 611-2408 611-2409 611-2410 611-2411 611-2399 611-5302 611-5303 611-5304 611-5305 611-5306 611-5307
Precision balances, Explorer®
Balances and Scales
A range of balances that incorporate a touch screen, advanced weighing and proximity sensor technologies. The easy to use control unit can be mounted on a base, above a base on an optional column, on a wall, or remotely (up to 3 metres distant with an optional cable). The touch screen display, with universal icons and informative prompts, simplifies set-up functions and basic operation. Then the operator has the added choice to use proximity sensors for hands-free operation when handling sensitive samples. These balances have multiple application modes from basic weighing to SQC applications, together with short stabilisation times and optimised performance specifications.
  • Colour touch screen display with viewing angle adjustment in a demountable control unit
  • Advanced weighing technology with stabilisation times of less than 2 seconds
  • SmarText™ software with text prompts for ease of menu navigation including 'level assist' feature for illuminated level indicator
  • AutoCal™ internal calibration
  • Dual multi-functional proximity sensors for hands-free operation of basic or user selectable functions
  • LIMS compatibility
1 mg balances feature a frameless glass draft shield with easy access, central light and removable doors for cleaning.
Ordering information: A selection of OIML EC approved balances are also available from Avantor. For the full range of accessories for the Explorer® please contact your local Avantor sales office.