Zinc, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
31653-1KG
31653-250G
SIAL31653-1KGEA
224
EUR
SIAL31653-1KG
SIAL31653-250G
Zinc, Sigma-Aldrich®
Zinc
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...