Analytical balances, Explorer®
Supplier: OHAUS
83021332 83021331 83021333 83021335 83021336
611-2394EA 5195 EUR
611-2394 611-2393 611-2395 611-2396 611-2397
Analytical balances, Explorer®
Balances and Scales
A range of four-place balances that incorporate a touch screen, advanced weighing and proximity sensor technologies. The easy to use control unit can be mounted on a base, above a base on an optional column, on a wall, or remotely (up to 3 metres distance with an optional cable). The touch screen display, with universal icons and informative prompts, simplifies set-up functions and basic operation. The operator has the added choice to use proximity sensors for hands-free operation when handling sensitive samples. These balances have multiple application modes from basic weighing to SQC applications, together with short stabilisation times and optimised performance specifications.
  • Colour touch screen display with viewing angle adjustment in a demountable control unit
  • Advanced weighing technology with stabilisation times of less than 3 seconds
  • SmarText™ software with text prompts for ease of menu navigation including 'Level assist' feature with illuminated level indicator
  • AutoCal™ internal calibration
  • Dual-multifunctional proximity sensors for hands-free operation of basic or user-selectable functions
  • LIMS compatibility
The balances feature a frameless glass draft shield with easy access, central light and removable doors for cleaning.
Ordering information: A selection of OIML EC approved balances are also available from Avantor. For models with automatic draft shield door operation via touch-free sensors and also a full range of accessories for the Explorer® balances, please contact your local Avantor sales office.