Chromium ≥99.2% (metals basis), pieces 3-12 mm (0.12-0.47 in)
Supplier: Thermo Fisher Scientific
00472.22 00472.36
00472.22EA 114 EUR
00472.22 00472.36
Chromium ≥99.2% (metals basis), pieces 3-12 mm (0.12-0.47 in)
Chromium
Purity: 99.2% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944
Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288