Chromium ≥99% (metals basis), irregular pieces
Supplier: Thermo Fisher Scientific
10150.22 10150.36
10150.22EA 85.9 EUR
10150.22 10150.36
Chromium ≥99% (metals basis), irregular pieces
Chromium
Purity: 99% CAS No.: 7440-47-3 Physical Form: Broken plate Odour: Odourless MDL No.: MFCD00010944
Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288