Chromium ≥99.996% (metals basis), powder -10+20 mesh
Supplier: Thermo Fisher Scientific
10452.06 10452.14 10452.22
10452.06EA 92.1 EUR
10452.06 10452.14 10452.22
Chromium ≥99.996% (metals basis), powder -10+20 mesh
Chromium
Purity: 99.996% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944
Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,02288