Chromium ≥99.99% (metals basis), powder -100+325 mesh
Supplier: Thermo Fisher Scientific
Warning
35668.09 35668.18
35668.09EA 145 EUR
35668.09 35668.18
Chromium ≥99.99% (metals basis), powder -100+325 mesh
Chromium
Purity: 99.99% CAS No.: 7440-47-3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00010944
Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,02288