Chromium ≥99.95% (metals basis), cubes typ. 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
42310.09 42310.18 42310.30
42310.09EA 56 EUR
42310.09 42310.18 42310.30
Chromium ≥99.95% (metals basis), cubes typ. 6.35 mm (0.25 in)
Chromium
Purity: 99.95% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944
Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288