Chromium ≥99.99% (metals basis), pieces 1-25 mm (0.04-1 in)
Supplier: Thermo Fisher Scientific
43066.A1 43066.30 43066.18
43066.A1EA 1720 EUR
43066.A1 43066.30 43066.18
Chromium ≥99.99% (metals basis), pieces 1-25 mm (0.04-1 in)
Chromium
Purity: 99.99% CAS No.: 7440-47-3 Physical Form: Broken plate Odour: Odourless MDL No.: MFCD00010944
Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288