Chromium ≥99.996% (metals basis), chunks 2-15 mm (0.08-0.59 in)
Supplier: Thermo Fisher Scientific
44565.09 44565.18
44565.09EA 111 EUR
44565.09 44565.18
Chromium ≥99.996% (metals basis), chunks 2-15 mm (0.08-0.59 in)
Chromium
Purity: 99.996% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944
Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288