Rhenium ≥99.997% (metals basis), powder, Puratronic® -22 mesh
Supplier: Thermo Fisher Scientific
Warning
10673.03 10673.06
10673.03EA 119 EUR
10673.03 10673.06
Rhenium ≥99.997% (metals basis), powder, Puratronic® -22 mesh
Rhenium
Purity: 99.997% CAS No.: 7440-15-5 UN No.: 3089 MDL No.: MFCD00011195
Formula: Re
MW: 186.21 g/mol
Boiling Pt: 5630
Melting Pt: 3180
Density: 21.02
Storage Temperature: Ambient
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,08343