Osmium 99.9% (trace metals basis)
Supplier: Thermo Fisher Scientific
Danger
385160010 385160050
ACRO385160010EA 168 EUR
ACRO385160010 ACRO385160050
Osmium 99.9% (trace metals basis)
Osmium
Formula: Os
Boiling Pt: 5027 °C (760 mmHg)
Melting Pt: 3045 °C
Storage Temperature: Ambient
MDL Number: MFCD00011147
CAS Number: 7440-04-2
EINECS: 231-114-0
UN: 3089
ADR: 4.1,II
Order Now
Specification Test Results
Appearance Blue-grey Powder
Particle Size/Sieve analysis ≤100 mesh
Total trace metal impurities ≤0.1 %