Cleanroom gloves, nitrile, sterile, Kimtech™ G3
Supplier: KIMBERLY CLARK
HC61160 HC61165 HC61170 HC61175 HC61180 HC61185
112-4837EA 0 EUR
112-4837 112-4838 112-4839 112-4840 112-4841 112-4842
Cleanroom gloves, nitrile, sterile, Kimtech™ G3
Gloves
Hand-specific, sterile gloves with beaded cuff for cleanroom applications.
  • Excellent dexterity and tactile sensitivity
  • Contains no natural rubber latex or silicone
  • Fully textured for improved grip
  • Low endotoxin levels
  • Validated to a Sterility Assurance Level (SAL) of 10⁻⁶
Suitable for use in Class 1, ISO 3 or M1.5 cleanrooms.
Length: 300 mm
Thickness: 0,13 mm (palm)
Colour: White
Certifications: PPE Cat III (EU) 2016/425, EN 374-1 Type C (K), EN 374-5 (VIRUS), AQL 1.5
Packaging: Walleted and pouched in polyethylene for cleanroom use. One pair/poly pouch, 20 pairs/double poly bag, 10 double bags (200 pairs)/case.