Hexane (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
227064-1L
227064-100ML
SIAL227064-1LEA
154
EUR
SIAL227064-1L
SIAL227064-100ML
Hexane (mixture of isomers), Sigma-Aldrich®
Hexane (mixture of isomers)
Formula:
H₃C(CH₂)₄CH₃ Boiling Pt: 68…70 °C (1013 hPa) Melting Pt: –95 °C Density: 0.66 g/cm³ (20 °C) Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179311 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...