Acetonitrile-[D₃]
Supplier: Roth Carl
Danger
Synonyms: Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]
AE52.1 AE52.2
ROTHAE52.1EA 103 EUR
ROTHAE52.1 ROTHAE52.2
Acetonitrile-[D₃]
Acetonitrile-[D3]
Formula: D₃CCN
Boiling Pt: 80.7 °C (1013 hPa)
Melting Pt: –42 °C
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II