Methanol-[D₄] 99.8 atom % D for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Danger
321280050 321280100 321280250 321281000
ACRO321280050EA 101 EUR
ACRO321280050 ACRO321280100 ACRO321280250 ACRO321281000
Methanol-[D₄] 99.8 atom % D for NMR spectroscopy
Methanol-[D4]
Formula: D₃COD
Boiling Pt: 65 °C (760 mmHg)
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
D-Enrichment ≥99.75 %