Nitrobenzene-[D₅] 99 atom % D for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Nitrobenzene-[²H₅]
166360050 166360100 166360250
ACRO166360050EA 58.4 EUR
ACRO166360050 ACRO166360100 ACRO166360250
Nitrobenzene-[D₅] 99 atom % D for NMR spectroscopy
Nitrobenzene-[D5]
Formula: C₆D₅NO₂
Boiling Pt: 88 °C (12 mmHg)
Melting Pt: 5 °C
Density: 1,25 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00044415
CAS Number: 4165-60-0
EINECS: 224-014-3
UN: 1662
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Water ≤0.03 %
D-Enrichment ≥98.5 %
HPLC ≥96.0 %