Trimethylamine 50% (w/w) 7.3 M in aqueous solution, pure
Supplier: Thermo Fisher Scientific
Danger
370785000 370780010 370780025 370780100
ACRO370785000EA 38.9 EUR
ACRO370785000 ACRO370780010 ACRO370780025 ACRO370780100
Trimethylamine 50% (w/w) 7.3 M in aqueous solution, pure
Trimethylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 30 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.86 g/cm³
Flash Pt: –45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 1297
ADR: 3 I
Merck Index: 12,09841
Order Now
Specification Test Results
Appearance Clear colorless Solution
Titration with HCl 48 to 52 %