Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
05030-1L-F 05030-2.5L-F 05030-500ML-F
SIAL05030-1L-FEA 267 EUR
SIAL05030-1L-F SIAL05030-2.5L-F SIAL05030-500ML-F
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution, Sigma-Aldrich®
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM