Bottles, narrow neck, Boston type, round, PPCO, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
312006-0016 312006-0032 312006-0002 312006-0001 312006-0004 312006-0008
215-2572EA 483 EUR
215-2572 215-2573 215-2568 215-2564 215-2569 215-2570
Bottles, narrow neck, Boston type, round, PPCO, with screw caps, Nalgene®
Bottles Plastic Bottles
PPCO, translucent with PP screw cap.
  • Excellent chemical resistance
  • Heat resistant up to +121 °C, autoclavable with or without contents
  • Robust, leakproof
Ideal for storing and shipping liquids.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904