Hydrochloric acid 35%, TECHNICAL
Supplier: VWR Chemicals
Danger
Synonyms: Hydrochloric acid solution, Muriatic acid
20246.298EA 18.3 EUR
20246.298 20246.323 20246.367 20246.460
Hydrochloric acid 35%, TECHNICAL
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
REACH: 01-2119484862-27
Order Now
Specification Test Results
Assay Min. 35 %