Orthophosphoric acid 85%
Supplier: Roth Carl
Danger
Synonyms: Phosphoric acid
6366.1 6366.2
ROTH6366.1EA 0 EUR
ROTH6366.1 ROTH6366.2
Orthophosphoric acid 85%
Orthophosphoric acid
Formula: H₃PO₄
Melting Pt: 28 °C
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III