(±)-2-Octanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
74858-100ML 74858-25ML
SIAL74858-100MLEA 422 EUR
SIAL74858-100ML SIAL74858-25ML
(±)-2-Octanol, Sigma-Aldrich®
(±)-2-Octanol
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 180
Melting Pt: –39 °C
Density: 0.818 g/cm³ (20 °C)
Flash Pt: 55
Storage Temperature: Ambient
MDL Number: MFCD00004591
CAS Number: 123-96-6
EINECS: 204-667-0
UN: 1987
ADR: 3 III
Merck Index: 13,06785