DL-Camphor
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone
JFD03952.250
JFD03952.1
MAYBJFD03952.250EA
0
EUR
MAYBJFD03952.250
MAYBJFD03952.1
DL-Camphor
DL-Camphor
Formula:
C₁₀H₁₆O Boiling Pt: 204 °C (1013 hPa) Melting Pt: ∼180 °C Density: 0.992 g/cm³ (25 °C) Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074738 CAS Number: 76-22-2 EINECS: 200-945-0 UN: 2717 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...