Sodium hydroxide 5 mol/l (5 N) concentrated aqueous solution, CLININORM
Supplier: VWR Chemicals
Danger
31625.293EA 63.3 EUR
31625.293 31625.362
Sodium hydroxide 5 mol/l (5 N) concentrated aqueous solution, CLININORM
Sodium hydroxide
CREUTZFELD-JAKOB disease
Dilute 1 volume in 4 volumes of water
Contains Bromothymol blue indicator
Formula: NaOH
Boiling Pt: >105 °C (1013 hPa)
Density: 1.327 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Titer 4.95 - 5.05 mol/l
Identification Passes test
NIST traceable Confirmed