Hydrochloric acid standardised solution 0.01 N
Supplier: Thermo Fisher Scientific
Synonyms: Hydrochloric acid solution, Muriatic acid
35648.K2 35648.M4
35648.K2EA 38.9 EUR
35648.K2 35648.M4
Hydrochloric acid standardised solution 0.01 N
Hydrochloric acid
CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colourless
Formula: HCl
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III