Sodium hydroxide standard solution 1 N in water
Supplier: Thermo Fisher Scientific
Danger
124260010 124260050 124260250
ACRO124260010EA 54.7 EUR
ACRO124260010 ACRO124260050 ACRO124260250
Sodium hydroxide standard solution 1 N in water
Sodium hydroxide
Formula: NaOH
Density: 1.04 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 0.998 to 1.002 N (20°C)