Stoppers for conical ground joints
Supplier: Brand
144620 144645 144640 144635 144625 144615 144610 144605 144630 144671 144672 144673 144674 144676
BRND144620EA 4.9 EUR
BRND144620 BRND144645 BRND144640 BRND144635 BRND144625 BRND144615 BRND144610 BRND144605 BRND144630 BRND144671 BRND144672 BRND144673 BRND144674 BRND144676
Stoppers for conical ground joints
Stoppers Glass Stoppers
DURAN®. Hollow, with drip tip and hexagonal grip.
Certifications: DIN 12252.