Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Supplier: Roth Carl
K022.1 K022.2
ROTHK022.1EA 24.7 EUR
ROTHK022.1 ROTHK022.2
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5