Sulphuric acid 0.02 mol/l (0.04 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
30146.291EA 20 EUR
30146.291 30146.400
Sulphuric acid 0.02 mol/l (0.04 N) in aqueous solution, AVS TITRINORM volumetric solution
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.004 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.01996 - 0.02004 mol/l
NIST traceable Confirmed