Sulphuric acid 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
30150.295EA 33.6 EUR
30150.295 30150.375 30150.320 30150.400
Sulphuric acid 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Sulphuric acid
Formula: H₂SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed