Sulphuric acid solution_standard 0.5 mol/l (1 N) water
Supplier: Thermo Fisher Scientific
Warning
124240010 124240025
ACRO124240010EA 49 EUR
ACRO124240010 ACRO124240025
Sulphuric acid solution_standard 0.5 mol/l (1 N) water
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 0.998 to 1.002 N (20°C)