Sulphuric acid 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
|
Warning
|
30144.294EA
37.6
EUR
30144.294
30144.328
30144.363
30144.408
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Sulphuric acid
Formula:
H₂SO₄ Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 3264 ADR: 8,III |
Specification Test Results
NIST traceable | Confirmed |
Titer (20°C; real value 0.2 % accuracy) | 0.499 - 0.501 mol/l |