Sulphuric acid 1 mol/l (2 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Warning
30149.291EA 45.8 EUR
30149.291 30149.325 30149.371
Sulphuric acid 1 mol/l (2 N) in aqueous solution, AVS TITRINORM volumetric solution
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
NIST traceable Confirmed