Sulphuric acid 2 mol/l (4 N) aqueous solution, AVS TITRINORM Volumetric
Supplier: VWR Chemicals
Danger
198156FEA 207 EUR
198156F 198154D
Sulphuric acid 2 mol/l (4 N) aqueous solution, AVS TITRINORM Volumetric
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.146 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 1.998 - 2.002 mol/l
NIST traceable Confirmed