Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Warning
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
30475.297EA 75.1 EUR
30475.297 30475.322
Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9 III
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed