Silver nitrate solution_standard 0.1 mol/l (0.1 N) water
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
124250010 124250050
ACRO124250010EA 107 EUR
ACRO124250010 ACRO124250050
Silver nitrate solution_standard 0.1 mol/l (0.1 N) water
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 1.015 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8 III
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 0.0998 to 0.1002 N (20°C)