Silver nitrate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Danger
30471.237EA 111 EUR
30471.237 30471.294
Silver nitrate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Silver nitrate
Formula: AgNO₃
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
NIST traceable Confirmed