Citric acid monohydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
M-Clarity™ quality level = MQ200
33114-1KG 33114-2.5KG 33114-25KG 33114-500G 33114-5KG 33114-6X1KG 33114-6X500G
SIAL33114-1KGEA 119 EUR
SIAL33114-1KG SIAL33114-2.5KG SIAL33114-25KG SIAL33114-500G SIAL33114-5KG SIAL33114-6X1KG SIAL33114-6X500G
Citric acid monohydrate, Sigma-Aldrich®
Citric acid monohydrate
Formula: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Melting Pt: 135…152 °C
Density: 1.552 g/cm³ (20 °C)
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM