Calcium chloride dihydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Calcium Chloride Dihydrate, Calcium dichloride
M-Clarity™ quality level = MQ200
21102-1KG-F 21102-25KG-F
SIAL21102-1KG-FEA 55.7 EUR
SIAL21102-1KG-F SIAL21102-25KG-F
Calcium chloride dihydrate, Sigma-Aldrich®
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660